missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GFAP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23935-1-AP-20UL
This item is not returnable.
View return policy
Description
GFAP (Glial fibrillary acidic protein) is a member of the class III intermediate filament protein family. GFAP is heavily, and specifically, expressed in astrocytes and certain astroglia in the central nervous system, in satellite cells in peripheral ganglia, and in non-myelinating Schwann cells in peripheral nerves. In addition, neural stem cells strongly express GFAP. Antibodies to GFAP are very useful as markers of astrocytic cells. In addition, many types of brain tumor, presumably derived from astrocytic cells, heavily express GFAP. GFAP is also found in the lens epithelium, Kupffer cells of the liver, in some cells in salivary tumors and has been reported in erythrocytes. GFAP is used as a marker to distinguish astrocytes from other glial cells during development. Mutations in this gene cause Alexander disease, a rare disorder of astrocytes in the central nervous system. Alternative splicing of the GFAP gene results in multiple transcript variants encoding distinct isoforms.Specifications
GFAP | |
Polyclonal | |
Unconjugated | |
GFAP | |
GFAP | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14580, 24387, 2670 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunofluorescence, Immunoprecipitation, Immunocytochemistry, Western Blot | |
0.41 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P03995, P14136, P47819 | |
Gfap | |
GFAP Fusion Protein Ag20853 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |