Learn More
Gellan Gum Powder, MP Biomedicals™
Catalog Number p-7083173
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
71010-52-1
(C24H38O20)A
Agar substitute gelling agent
71010-52-1
(C24H38O20)A
Agar substitute gelling agent
Used as a gelling agent in place of agar in microbiological media. Gellan Gum Powder, MP Biomedicals is a water-soluble polysaccharide produced by Pseudomonas elodea, a bacterium. It is used in a variety of immobilization matricies and as a food additive,
Quantity 100 g
Physical Form Powder