missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FUT3 Mouse anti-Human, Unconjugated, Clone: 2A6G7, Proteintech
Mouse Monoclonal Antibody
140.00 EUR - 399.00 EUR
Specifications
Antigen | FUT3 |
---|---|
Clone | 2A6G7 |
Concentration | 1000 μg/mL |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence, ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16869925
|
Proteintech
67344-1-IG-20UL |
20 μL |
140.00 EUR
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16859925
|
Proteintech
67344-1-IG-150UL |
150 μL |
399.00 EUR
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The FUT3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human FUT3. This antibody recognizes human antigen. The FUT3 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
FUT3 | |
1000 μg/mL | |
Monoclonal | |
Mouse | |
2525 | |
Primary |
2A6G7 | |
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
Unconjugated | |
Human | |
IgG1 |