Learn More
Poulten Graf™ Fortuna™ Optifix™ Safety Dispenser
For dispensing of strong acids and aggressive chemicals (no HF). Poulten Graf™ Fortuna™ Optifix™ Safety Dispensers have a dispensing cylinder made of glass and a PTFE-encased piston with sealing lip.
Brand:  Poulten Graf™ 101 10733S
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 10341222
  • 651.00 EUR / Each
Estimated Shipment: 14-06-2024
Description
For dispensing of strong acids and aggressive chemicals (no HF). Poulten Graf™ Fortuna™ Optifix™ Safety Dispensers have a dispensing cylinder made of glass and a PTFE-encased piston with sealing lip.
  • Manufactured according to DIN EN ISO 9001:2008 using high quality materials
  • With certificate, individually numbered
  • Glass dispensing cylinder is coated with plastic to provide breakage protection
  • Ergonomic finger protection cap made of PP with volume setting system
  • Graduation on the scale is lasered and easy to read
  • Fast and easy volume setting with additional fine adjustment
Specifications
Specifications
Outfix Safety
1 to 5 mL
PTFE-coated Piston, Stopcock and Ventilator Valve