Learn More
Proteintech FlexAble FITC Plus Antibody Labeling Kit for Human IgG
FlexAble FITC Plus Antibody Labeling Kit for Human IgG is a novel antibody labeling kit that uses an affinity linker to conjugate FITC Plus in any buffer condition to Human IgG primary antibodies from any supplier.
107.31 EUR - 1059.97 EUR
Specifications
Format Liquid
View More Specs
Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18085155
View Documents Promotion Details
Proteintech
KFA109-10
10 Tests
107.31 EUR
10 reactions
View alternative products
Save Up to 
»
18030596
View Documents Promotion Details
Proteintech
KFA109-200
100 Tests
1059.97 EUR
200 reactions
View alternative products
Save Up to 
»
18007295
View Documents Promotion Details
Proteintech
KFA109-50
50 Tests
499.58 EUR
50 reactions
View alternative products
Save Up to 
»
Description
Description
FlexAble FITC Plus Antibody Labeling Kit for Human IgG is a novel antibody labeling kit that uses an affinity linker to conjugate FITC Plus in any buffer condition to Human IgG primary antibodies from any supplier. One labeling reaction requires only 0.5 μg of antibody - regardless of the antibody concentration. Label your Human IgG antibody in<10 min without any additional equipment.