Learn More
Fisherbrand™ Rotary Mixer Discs
Suitable for end-to-end mixing tubes
122.00 EUR
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
12377488
|
Fisherbrand™
|
Tubes with a Diameter Between 8-11 mm |
122.00 EUR
Each |
Estimated Shipment: 10-07-2024
Log in to see stock. |
|||||
12387488
|
Fisherbrand™
|
Tubes with a Diameter Between 15-17 mm |
122.00 EUR
Each |
Estimated Shipment: 16-09-2024
Log in to see stock. |
|||||
Description
- Rotary Mixer Disc complete with clips for tubes, supplied unassembled
- Suitable for 'End to End Mixing' tubes,
- Clips for tube diameters between 8 - 11 mm are suitable for 0.5mL, 1.5mL and 2.0mL Microcentrifuge tubes, 3mL Vacutainers and 5mL capped tubes (8 -11 mm)
- Clips for tube diameters between 15 - 17 mm are suitable for 15 mL centrifuge tubes
- Easy to fit onto the RM-1 locating on three pins then secured in place with thumb wheel
Specifications
Rotary Mixer Disc |