Learn More
Fisherbrand™ Multipurpose Specimen Storage Containers
Inert to most chemicals including formaldehyde, weak acids and all bases
12.55 EUR - 96.40 EUR
Product Code | Brand | Capacity (Metric) | Closure Material | Color | Shape | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Closure Material | Color | Shape | Price | Quantity & Availability | ||
12029977
|
Fisherbrand™
|
118 mL | Polypropylene | Translucent | Short, Wide |
45.65 EUR
Pack of 300 |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11709388
|
Fisherbrand™
|
148 mL | Polypropylene | Translucent | Tall, Thin |
96.40 EUR
Pack of 300 |
Estimated Shipment: 21-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
12049977
|
Fisherbrand™
|
237 mL | LDPE | Translucent | Short, Wide |
17.90 EUR
Pack of 100 |
Estimated Shipment: 03-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11719388
|
Fisherbrand™
|
237 mL | LDPE | White | Short, Wide |
41.25 EUR
Pack of 100 |
Estimated Shipment: 21-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11974375
|
Fisherbrand™
|
237 mL | LDPE | Translucent | Tall, Thin |
33.40 EUR
Pack of 100 |
Estimated Shipment: 05-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
12029957
|
Fisherbrand™
|
237 mL | LDPE | White | Tall, Thin |
21.00 EUR
Pack of 100 |
Estimated Shipment: 16-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
12089977
|
Fisherbrand™
|
473 mL | LDPE | Translucent | Short, Wide |
23.75 EUR
Pack of 100 |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11994375
|
Fisherbrand™
|
473 mL | LDPE | White | Tall, Thin |
50.85 EUR
Pack of 100 |
Estimated Shipment: 05-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
12089947
|
Fisherbrand™
|
946 mL | LDPE | Tall, Thin |
36.70 EUR
Pack of 100 |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12089957
|
Fisherbrand™
|
1100 mL | LDPE | White | Short, Wide |
40.05 EUR
Pack of 100 |
Estimated Shipment: 23-05-2024
Log in to see stock. |
||
12009957
|
Fisherbrand™
|
1900 mL | LDPE | Translucent | Tall, Thin |
35.55 EUR
Pack of 50 |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
12009967
|
Fisherbrand™
|
1900 mL | LDPE | White | Tall, Thin |
32.55 EUR
Pack of 50 |
Estimated Shipment: 03-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
12039957
|
Fisherbrand™
|
2500 mL | LDPE | Tall, Thin |
18.45 EUR
Pack of 25 |
Estimated Shipment: 23-05-2024
Log in to see stock. |
|||
12069967
|
Fisherbrand™
|
2500 mL | LDPE | White | Tall, Thin |
19.10 EUR
Pack of 25 |
Estimated Shipment: 05-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
12099957
|
Fisherbrand™
|
5100 mL | LDPE | Translucent | Short, Wide |
13.50 EUR
Pack of 10 |
Estimated Shipment: 23-09-2024
Log in to see stock. |
||
12089967
|
Fisherbrand™
|
5100 mL | LDPE | White | Tall, Thin |
12.55 EUR
Pack of 10 |
Estimated Shipment: 05-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
Description
Fisherbrand™ Multipurpose Specimen Containers are designed for collection, transport, and storage of liquid and dry specimens and samples; nonsterile. Inert to most chemicals including formaldehyde, weak acids and all bases.
- Resist freezing and boiling
- Stackable for compact storage
- Select containers are autoclavable (see ordering table); lids are not autoclavable
1 Short/wide style
2 Tall/thin style
3 Graduated
Specifications
Specimen Container |