Learn More
Fisherbrand™ Filter Cards for Shandon™ Cytospin™
White cards are made from a blend of 100% cotton and cellulose fibers
Brand: Fisherbrand™
Additional Details : Weight : 0.18900kg
Description
White cards are made from a blend of 100% cotton and cellulose fibers
- Excellent absorbent padding for the collection of excess reagents or wash solution
- Cellulose fibers improve the card's strength and durability during centrifugation and facilitate the removal of the filter card from the slide
Specifications
Shandon Cytospin | |
Filter Card |