Learn More
Fisherbrand™ Replacement Pouches for FB-GC20-1 Vertical Gel Caster
Replacement Pouches for FB-GC20-1 Vertical Gel Caster
21.30 EUR - 48.60 EUR
Specifications
For Use With (Equipment) | Owl™ JGC-3 Gel Casting System |
---|---|
Type | Gel Pouch |
Gel Dimensions | 20 x 20 cm |
Length (Metric) | 20 cm |
Width (Metric) | 20 cm |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12424647
|
Fisherbrand™
|
25/Pk. |
21.30 EUR
Pack of 25 |
Estimated Shipment: 23-09-2024
Log in to see stock. |
|||||
11972975
|
Fisherbrand™
|
100/Pk. |
48.60 EUR
Pack of 100 |
Estimated Shipment: 17-09-2024
Log in to see stock. |
|||||
Specifications
Owl™ JGC-3 Gel Casting System | |
20 x 20 cm | |
20 cm |
Gel Pouch | |
20 cm |