Learn More
Thermo Scientific™ Finntip™ Stepper Pipette Tips
Integrate Thermo ScientificTM FinntipTM Stepper Pipette Tips with the Thermo ScientificTM FinnpipetteTM Stepper Pipetter.
19.90 EUR - 193.00 EUR
Specifications
Filtered | Non-Filtered |
---|---|
Certifications/Compliance | ISO 9001, ISO 14001 |
For Use With (Equipment) | Thermo Scientific FinnPipette Stepper |
Product Line | FinnTip |
Product Code | Brand | Volume (Metric) | Sterility | Volume (Metric) Dispensing | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Sterility | Volume (Metric) Dispensing | Price | Quantity & Availability | |||
10626111
|
Thermo Scientific™
9404170 |
500 μL | Non-sterile | 10, 20, 30, 40, 50μL |
193.00 EUR
Pack of 100 |
In Stock
Log in to see stock. |
|||
10411891
|
Thermo Scientific™
9404173 |
500 μL | Sterile | 10, 20, 30, 40, 50μL |
146.00 EUR
Pack of 50 |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||
10185900
|
Thermo Scientific™
9404180 |
1250 μL | Non-sterile | 25, 50, 75, 100, 125μL |
137.00 EUR
Pack of 100 |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||
11782574
|
Thermo Scientific™
9404183 |
1250 μL | Sterile | 25, 50, 75, 100, 125μL |
138.00 EUR
Pack of 50 |
In Stock
Log in to see stock. |
|||
10542481
|
Thermo Scientific™
9404190 |
2.5 mL | Non-sterile | 50, 100, 150, 200, 250μL |
137.00 EUR
Pack of 100 |
In Stock
Log in to see stock. |
|||
11792574
|
Thermo Scientific™
9404193 |
2.5 mL | Sterile | 50, 100, 150, 200, 250μL |
138.00 EUR
Pack of 50 |
In Stock
Log in to see stock. |
|||
10762111
|
Thermo Scientific™
9404200 |
5 mL | Non-sterile | 100, 200, 300, 400, 500μL |
104.00 EUR
Pack of 50 |
In Stock
Log in to see stock. |
|||
11702584
|
Thermo Scientific™
9404203 |
5 mL | Sterile | 100, 200, 300, 400, 500μL |
69.25 EUR
Pack of 25 |
In Stock
Log in to see stock. |
|||
11742574
|
Thermo Scientific™
9404210 |
12.5 mL | Non-sterile | 250, 500, 750, 1000, 1250μL |
67.70 EUR
Pack of 50 |
In Stock
Log in to see stock. |
|||
11712584
|
Thermo Scientific™
9404213 |
12.5 mL | Sterile | 250, 500, 750, 1000, 1250μL |
68.85 EUR
Pack of 25 |
In Stock
Log in to see stock. |
|||
10401891
|
Thermo Scientific™
9404220 |
25 mL | Non-sterile | 500, 1000, 1500, 2000, 2500μL |
35.90 EUR
Pack of 20 |
In Stock
Log in to see stock. |
|||
11722584
|
Thermo Scientific™
9404223 |
25 mL | Sterile | 500, 1000, 1500, 2000, 2500μL |
36.00 EUR
Pack of 10 |
In Stock
Log in to see stock. |
|||
10083630
|
Thermo Scientific™
9404230 |
50 mL | Non-sterile | 1000, 2000, 3000, 4000, 5000μL |
19.90 EUR
Pack of 10 |
In Stock
Log in to see stock. |
|||
10750981
|
Thermo Scientific™
9404233 |
50 mL | Sterile | 1000, 2000, 3000, 4000, 5000μL |
54.05 EUR
Pack of 10 |
In Stock
Log in to see stock. |
|||
Description
Integrate Thermo Scientific™ Finntip™ Stepper Pipette Tips with the Thermo Scientific™ Finnpipette™ Stepper Pipetter.
- Nonsterile tips are bulk packaged; sterile tips are individually wrapped
Specifications
Non-Filtered | |
ISO 9001, ISO 14001 | |
Thermo Scientific FinnPipette Stepper | |
FinnTip |