Learn More
Agilent BioTek Filter Cube
For use with Synergy LX
Brand:  Agilent BioTek 1505004
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 15978550
  • 1025.00 EUR / 1 unit
Estimated Shipment: 14-08-2024
Specifications
Synergy LX
EX 530/25, EM 590/25, 570 nm mirror (Red)