missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferritin, Equine, MP Biomedicals™
Click to view available options
Quantity:
100 mg
500 mg
Description
A shell of 24 protein subunits (apoferritin) and a core of Fe3+ ions with a peptide molecular weight of 440 kDa. The ferritin-like superfamily of iron-storage proteins is virtually ubiquitous in life on earth.
- Studies have shown that ferritin iron incorporation is mediated by a ferroxidase activity associated with ferritin H subunits and a nucleation center associated with ferritin L subunits.
- Release of iron from ferritin has an essential role in iron-dependent lipid peroxidation and may contribute to free radical-induced cell damage in vivo.
- Therefore, by binding iron, ferritin may function as an antioxidant.
Specifications
Quantity | 100 mg |
Solubility Information | Soluble in 0.15M sodium chloride, distilled water or dilute buffer. |
Formula Weight | ∼450,000 (Lit.) |
Physical Form | Viscous Liquid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'