missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Etoposide, MP Biomedicals
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Specifications
Color | White |
Quantity | 25 mg |
IUPAC Name | (10R,11R,15R,16S)-16-{[(2R,4aR,6R,7R,8R,8aS)-7,8-dihydroxy-2-methyl-hexahydro-2H-pyrano[3,2-d][1,3]dioxin-6-yl]oxy}-10-(4-hydroxy-3,5-dimethoxyphenyl)-4,6,13-trioxatetracyclo[7.7.0.0³,⁷.0¹¹,¹⁵]hexadeca-1,3(7),8-trien-12-one |
Formula Weight | 588.6 |
Physical Form | Powder |
Chemical Name or Material | Etoposide |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS07_Warning.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS08_Dangerous_Health_Hazard.png)
Product Identifier
- Etoposide
- Danger
- Acute toxicity Category 4
- Carcinogenicity Category 1B
- Reproductive toxicity Category 2
- H302-Harmful if swallowed.
- H350-May cause cancer.
- H361-Suspected of damaging fertility or the unborn child.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : etoposide
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'