Learn More
DWK Life Sciences DURAN™ Weighing Bottle, with ground lid, low form
Weighing glasses are suitable for weighing substances. The matching lid prevents unintentional distribution of the weighing product.
312.00 EUR - 550.00 EUR
Specifications
Description | Low form |
---|---|
For Use With (Application) | Weighing; Storage and transport; Available in low and high forms |
Material | Borosilicate Glass (3.3) |
Quantity | 10/Pk. |
Autoclavable | Autoclavable |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
11730944
|
DWK Life Sciences
242101304 |
5 mL |
312.00 EUR
Pack of 10 |
Estimated Shipment: 29-10-2024
Log in to see stock. |
|||||
11740944
|
DWK Life Sciences
242102309 |
15 mL |
354.00 EUR
Pack of 10 |
Estimated Shipment: 01-11-2024
Log in to see stock. |
|||||
11700964
|
DWK Life Sciences
242103202 |
30 mL |
400.00 EUR
Pack of 10 |
Estimated Shipment: 29-10-2024
Log in to see stock. |
|||||
11710964
|
DWK Life Sciences
242104104 |
80 mL |
550.00 EUR
Pack of 10 |
Estimated Shipment: 29-10-2024
Log in to see stock. |
|||||
Description
In order to carry out reactions in a controlled manner, it is essential to know the quantities of substances required and to weigh the corresponding substances accurately. A weighing glass can be used for weighing. The associated lid allows the weighing pan to be closed and thus prevents, for example, turbulence of the weighed-in solids during transport. Since even the smallest contamination of the weighing glass can have an effect on the weighing inheritance, the glass in the area of the ground joint and the lid knob are so pronounced that they can be grasped and moved with forceps or tweezers.
- Close-fitting lid prevents any sample loss during transport after weighing
- Fired-on, highly durable white ceramic
Specifications
Low form | |
Borosilicate Glass (3.3) | |
Autoclavable |
Weighing; Storage and transport; Available in low and high forms | |
10/Pk. |