Learn More
DWK Life Sciences DURAN™ Filter Disc Sintered Glass, Porosity 5, edge not fused
A prerequisite for successful work with glass filters is the selection of the right porosity. It is important to select a filter whose largest pore size is slightly below the grain size of the smallest particles still to be separated.
58.05 EUR - 359.00 EUR
Specifications
Material | Borosilicate Glass (3.3) |
---|---|
For Use With (Application) | Suitable for vacuum filtration with a maximum resulting pressure difference of 1 bar |
Product Code | Brand | Diameter (Metric)Top | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric)Top | Price | Quantity & Availability | |||||
11689998
|
DWK Life Sciences
251555502 |
55 mm |
58.05 EUR
Each |
Estimated Shipment: 23-07-2024
Log in to see stock. |
|||||
11699998
|
DWK Life Sciences
251560505 |
60 mm |
64.75 EUR
Each |
Estimated Shipment: 23-08-2024
Log in to see stock. |
|||||
11680009
|
DWK Life Sciences
251590508 |
90 mm |
119.00 EUR
Each |
Estimated Shipment: 23-08-2024
Log in to see stock. |
|||||
11640019
|
DWK Life Sciences
251600506 |
100 mm |
199.00 EUR
Each |
Estimated Shipment: 23-07-2024
Log in to see stock. |
|||||
11629998
|
DWK Life Sciences
251550504 |
50 mm |
183.00 EUR
Pack of 5 |
Estimated Shipment: 23-08-2024
Log in to see stock. |
|||||
11619988
|
DWK Life Sciences
251525508 |
25 mm |
229.00 EUR
Pack of 10 |
Estimated Shipment: 23-07-2024
Log in to see stock. |
|||||
11659988
|
DWK Life Sciences
251540503 |
40 mm |
322.00 EUR
Pack of 10 |
Estimated Shipment: 23-07-2024
Log in to see stock. |
|||||
11660009
|
DWK Life Sciences
251570506 |
70 mm |
81.40 EUR
Each |
Estimated Shipment: 23-07-2024
Log in to see stock. |
|||||
11649978
|
DWK Life Sciences
251510509 |
10 mm |
165.00 EUR
Pack of 10 |
Estimated Shipment: 23-07-2024
Log in to see stock. |
|||||
11649988
|
DWK Life Sciences
251535509 |
35 mm |
274.00 EUR
Pack of 10 |
Estimated Shipment: 23-07-2024
Log in to see stock. |
|||||
11650009
|
DWK Life Sciences
251565503 |
65 mm |
70.05 EUR
Each |
Estimated Shipment: 23-07-2024
Log in to see stock. |
|||||
11619998
|
DWK Life Sciences
251545501 |
45 mm |
359.00 EUR
Pack of 10 |
Estimated Shipment: 23-07-2024
Log in to see stock. |
|||||
11670009
|
DWK Life Sciences
251580507 |
80 mm |
99.45 EUR
Each |
Estimated Shipment: 23-07-2024
Log in to see stock. |
|||||
11650019
|
DWK Life Sciences
251620508 |
120 mm |
228.00 EUR
Each |
Estimated Shipment: 23-07-2024
Log in to see stock. |
|||||
11699978
|
DWK Life Sciences
251515507 |
15 mm |
168.00 EUR
Pack of 10 |
Estimated Shipment: 23-07-2024
Log in to see stock. |
|||||
11629988
|
DWK Life Sciences
251530502 |
30 mm |
249.00 EUR
Pack of 10 |
Estimated Shipment: 23-07-2024
Log in to see stock. |
|||||
11609988
|
DWK Life Sciences
251520501 |
20 mm |
168.00 EUR
Pack of 10 |
Estimated Shipment: 23-07-2024
Log in to see stock. |
|||||
Description
Glass powder with a certain particle size distribution results in a porous, solid body. These bodies are used as filter material in the form of plates and candles. Made of DURAN™ (borosilicate glass 3.3). They are ideally suited for separations with strong acids or alkalis, for example, and thus offer advantages over other materials such as plastic or paper. Available in 6 different porosity classes according to ISO 4796 as well as ASTM EI28-99. Suitable for vacuum filtration with a maximum resulting pressure difference of 1 bar.
- Edge non-melted and centered, surfaces untreated
- Successful work through the correct choice of porosity (The largest pore size should be slightly below the grain size of the smallest particles still to be separated).
- Available in 6 different porosity classes according to ISO 4796 as well as ASTM EI28-99.
- Glass articles for further processing
Specifications
Borosilicate Glass (3.3) | |
Suitable for vacuum filtration with a maximum resulting pressure difference of 1 bar |