Learn More
Fisherbrand™ Drosophila Products, BuzzPlugs™ Vial and Bottle Closures
Nonshedding, dense weave of nontoxic cellulose acetate minimizes evaporation, keeps food fresh longer
132.00 EUR - 147.00 EUR
Specifications
Material | Cellulose Acetate |
---|---|
Type | BuzzPlugs Bottle Closure |
For Use With (Application) | Minimizes evaporation, keeps food fresh longer |
Includes | Superior barrier |
Quantity | 1000 Case |
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
10346221
|
Fisherbrand™
|
2.5cm narrow plastic shell vials |
132.00 EUR
Pack of 1000 |
Estimated Shipment: 30-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10550152
|
Fisherbrand™
|
2.8cm wide plastic shell vials |
147.00 EUR
Pack of 1000 |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Nonshedding, dense weave of nontoxic cellulose acetate minimizes evaporation, keeps food fresh longer
- Superior barrier to mite infestation
- Exact fit with narrow 0.9 in. (25mm) or wide 1.1 in. (28.5mm) dia. plastic shell vials or standard (≈24mm dia.) glass vials or plastic stock bottles
- Stretches yet maintains tight fit after repeated removal and closing
- Flat surface accepts marking pen
Specifications
Cellulose Acetate | |
Minimizes evaporation, keeps food fresh longer | |
1000 Case |
BuzzPlugs Bottle Closure | |
Superior barrier |