Learn More
DRD1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17934-1-AP-150UL
This item is not returnable.
View return policy
Description
The D1 subtype is the most abundant dopamine receptor in the central nervous system. This G-protein coupled receptor stimulates adenylyl cyclase and activates cyclic AMP-dependent protein kinases. D1 receptors regulate neuronal growth and development, mediate some behavioral responses, and modulate dopamine receptor D2-mediated events. Alternate transcription initiation sites result in two transcript variants of this gene.Specifications
DRD1 | |
Polyclonal | |
Unconjugated | |
DRD1 | |
D(1A) dopamine receptor, DADR, Dopamine D1 receptor, dopamine receptor D1, DRD1, DRD1A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
13488, 1812 | |
-20°C | |
Liquid |
Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P21728, Q61616 | |
DRD1 | |
DRD1 Fusion Protein Ag12366 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |