Learn More
Miele Professional™ Dosing system
Bellows dispenser pump DOS NA 120 for dispensing liquid neutral/alkaline media (PG 8536)
Brand: Miele Professional™ 7514610
Additional Details : Weight : 2.94000kg
Description
- For dispensing liquid cleaning agent, emulgator
- For dispensing chemical disinfectant
- For retro-fitting by Miele Service
Specifications
Dispensing System | |
Washer-disinfectors, laboratory |