missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethylene Glycol Dimethyl Ether, 99%, Reagent Grade, Honeywell™
Click to view available options
Quantity:
5 mL
500 mL
1 L
2.5 L
20 L
Packaging:
Glass bottle
Stainless steel can
Description
- ≥95% for general laboratory use.
Specifications
Specifications
Melting Point | -64°C |
Color | Colorless |
Density | 0.944g/mL at 20°C, 0.939g/mL at 25°C |
Boiling Point | 162°C |
Flash Point | 57°C (134.6°F) |
Odor | Mild ether-like |
Packaging | Glass bottle |
Linear Formula | (CH3OCH2CH2)2O |
Quantity | 1 L |
UN Number | UN3271 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'