missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bollé Safety B-Clean Lens Cleaning Products/Kits
Click to view available options
Product Type:
Anti-Fog Kit
Anti-Fog Spray
Cleaning Station
Lens Cleaner Spray
Multi-function Dry Cleaning Tissues
Tissue Dispenser
Volume (Metric):
30 mL
50 mL
250 mL
500 mL
Description
- Alcohol and silicone free
- Water-based formula guarantees a perfect cleaning and protection for your spectacles and goggles
Specifications
Specifications
Product Type | Lens Cleaner Spray |
Description | Lens Cleaning Accessories |
For Use With (Application) | For All Types of Ocular |
Volume (Metric) | 250 mL |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'