Learn More
Groesfjeld Diagnostics deconex™ 22 PF
Developed for a wide range of automated applications. Groesfjeld Diagnostics deconex™ 22 PF is an alkaline cleaning powder used to remove organic and inorganic residues and contamination from glass and other reusable lab materials. This powder is free of NTA, phosphate, chlorine and surfactants.
POA
Specifications
Form | Powder |
---|---|
Appearance | White |
Composition | Alkalis, complexing agents, corrosion inhibitor |
Description | Suitable for stainless steel, laboratory glass, ceramics, and synthetic materials. Not suitable for aluminum, anodised aluminum, zinc, non-ferrous metals, rubber or latex. |
For Use With (Application) | Cleaning and decontamination |
Description
deconex™ 22 PF is a special cleaner that has been developed for a wide range of automated applications. The product has excellent dirt-removing properties, and it acts very effectively against heavy contamination and dried-on contaminants.
Due to its excellent dispersing properties, this product prevents redepositing of removed contaminations onto the cleaned parts. It provides completely residue-free cleaning of laboratory glass and reusable materials for successful work in analytical, synthesis and cell culture laboratories.
- Removes analytical residues, dried synthesis residues (like polymers or organic chemicals), organic and inorganic salts, greases, oils, pigments, blood, tissue residues and proteins
- Suitable for use in chemical, microbiological and medical laboratories, and in the pharmaceutical and food industries
- For neutralization of carried over alkaline residues, an acidic rinse should always follow the cleaning steps
- pH (1% solution in demineralized water): approx. 11.7–12.7
Specifications
Powder | |
Alkalis, complexing agents, corrosion inhibitor | |
Cleaning and decontamination |
White | |
Suitable for stainless steel, laboratory glass, ceramics, and synthetic materials. Not suitable for aluminum, anodised aluminum, zinc, non-ferrous metals, rubber or latex. |