Learn More
Cyclooctylamine, 97+%, Thermo Scientific™
23.40 EUR - 54.65 EUR
Chemical Identifiers
CAS | 5452-37-9 |
---|---|
Molecular Formula | C8H17N |
Molecular Weight (g/mol) | 127.231 |
MDL Number | MFCD00001748 |
InChI Key | HSOHBWMXECKEKV-UHFFFAOYSA-N |
Synonym | cyclooctylamine, aminocyclooctane, cyclooctyl amine, acmc-1ap2c, hsohbwmxeckekv-uhfffaoysa, cyclooctylamine, purum gc |
PubChem CID | 2903 |
IUPAC Name | cyclooctanamine |
SMILES | C1CCCC(CCC1)N |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsCyclooctylamine is used as pharmaceutical intermediates.
Solubility
Soluble in water. 10.2 g/L at 20°C.
Notes
Protect from moisture, heat. Avoid excessive and light exposure. Air sensitive. Store at +15°C to +25°C.
Chemical Identifiers
5452-37-9 | |
127.231 | |
HSOHBWMXECKEKV-UHFFFAOYSA-N | |
2903 | |
C1CCCC(CCC1)N |
C8H17N | |
MFCD00001748 | |
cyclooctylamine, aminocyclooctane, cyclooctyl amine, acmc-1ap2c, hsohbwmxeckekv-uhfffaoysa, cyclooctylamine, purum gc | |
cyclooctanamine |
Specifications
5452-37-9 | |
0.912 | |
62°C (143°F) | |
C8H17N | |
MFCD00001748 | |
2689120 | |
cyclooctylamine, aminocyclooctane, cyclooctyl amine, acmc-1ap2c, hsohbwmxeckekv-uhfffaoysa, cyclooctylamine, purum gc | |
HSOHBWMXECKEKV-UHFFFAOYSA-N | |
cyclooctanamine | |
2903 | |
≥97% |
-48°C | |
189°C to 190°C | |
Amine-like | |
1.481 | |
UN2735 | |
Air Sensitive | |
Soluble in water. 10.2g/L at 20°C. | |
C1CCCC(CCC1)N | |
127.231 | |
127.23 | |
Cyclooctylamine |
Safety and Handling
GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H227-H314-H335
missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: II
missing translation for 'einecsNumber' : 226-694-7
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Argon
- Cyclooctylamine
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.