Learn More
Ambion™ Custom Silencer™ siRNA
Custom Silencer™ siRNA
Brand: Ambion™ AM16230
845.54 EUR valid until 2024-06-28
Use promo code "21649" to get your promotional price.
Additional Details : Weight : 0.00450kg
Description
RNA interference (RNAi) is the best way to effectively knock down gene expression to study protein function in a wide range of cell types. Use our GeneAssist™ Custom siRNA Builder to design and order a custom siRNA.Specifications
Room temperature | |
Custom siRNA | |
100 nmol |
Custom siRNA, 100nmol, In-vivo-Purity, RNAi is the best Way to Effectively Knock Down Gene Expression to Study Protein Function in a Wide Range of Cell Types, Use our GeneAssist Custom siRNA Builder to Design and Order a Custom siRNA | |
In-vivo |
For Research Use Only. Not for use in diagnostic procedures.