Learn More
Corning™ Penicillin-Streptomycin Solution
Effective against Gram-positive and Gram-negative bacteria. Corning™ Penicillin-Streptomycin Solution is a mix of the antibiotics Penicillin (10,000 IU) and Streptomycin (10,000μg/mL) in a 100-fold working concentration.
Brand: Corning™ 30-002-CI
This item is not returnable.
View return policy
Description
- Certificate of Analysis available
- Penicillin (Penicillin G) works by inhibiting peptidoglycan synthesis, while Streptomycin inhibits protein synthesis
Specifications
-20°C | |
Solution | |
Sterile | |
6 x 100 mL |
10,000 IU Penicillin, 10,000μg/mL Streptomycin | |
Antibiotic | |
Plastic Bottle |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Penicillin-Streptomycin Solution
Signal Word
- Danger
Hazard Category
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
- Reproductive toxicity Category 2
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P363-Wash contaminated clothing before reuse.
- P501-Dispose of contents/container to .
Supplemental information
- MIXTURE LIST-Contains : Streptomycin sulfate, Penicillin G potassium salt