Learn More
Corn Oil, MP Biomedicals
Catalog Number p-3545919
Click to view available options
Quantity:
1 lb.
5 lb.
25 lb.
8001-30-7
8001-30-7
Oleic Acid: 43-47%; Linoleic Acid: 39-42%; Palmitic Acid: 7-8%; Stearic Acid: 3-4%, Arachidic Acid: 0.5%; Approx. 7.6 lbs/gal
Quantity 1 lb.
Chemical Name or Material Corn Oil