Learn More
Epredia™ ColorStack Storage System
Take organization of your slides archives one step further with color-coded drawers.
Brand: Epredia™ E43/12C/AQUA
Additional Details : Weight : 12.00000kg
Description
- Fully compatible with all standard cabinets, bases, and options
- Cabinet is cream epoxy-coated metal; drawers have colored plastic fronts
- Each cabinet comes with 14 drawers of one color
- Drawers in the standard 14-Drawer Cabinet can easily be switched out with the ColorStack drawers
- Special top cover is cream to match the ColorStack 14-Drawer cabinet color
Specifications
14-Drawer Cabinet | |
Aqua |