Learn More
Tenak™ Color Inserts for SlimTube™ Cryogenic Vials
Provide quick and easy sample identification. Tenak™ Color Inserts for SlimTube™ Cryogenic Vials fit snugly in the top of the SlimTube cap.
29.13 EUR - 98.10 EUR
Product Code | Brand | Color | No. per Pack | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | No. per Pack | Price | Quantity & Availability | ||||
15605029
|
Tenak™
TE78400 |
MonoColor Red | 500 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15615029
|
Tenak™
TE78405 |
MonoColor Blue | 500 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15635029
|
Tenak™
TE78415 |
MonoColor Green | 500 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15645029
|
Tenak™
TE78420 |
MonoColor Purple | 500 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15655029
|
Tenak™
TE78425 |
MonoColor White | 500 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15625029
|
Tenak™
TE78410 |
MonoColor Yellow | 500 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15695019
|
Tenak™
TE78365 |
Assorted DualColors: Purple/White, Yellow/Green, Red/Blue | 600 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Available in MonoColor (one color) or DualColor (two different colored sides)
- Packaged in bags of 100
- For use with SlimTube Cryogenic Vials
Specifications
SlimTube cryogenic vials | |
Round |