Learn More
Masterflex™ Male Luer Fitting with Lock Ring
Use these luer fittings for a variety of applications, including syringes or small tubing. Cole-Parmer™ Male Luer Fittings with Lock Ring feature full material traceability to ensure process integrity.
Brand:  Masterflex™ 45518-06
Additional Details : Weight : 0.01530kg
Product Code. 15164542
  • 22.80 EUR / Pack of 25
Estimated Shipment: 23-07-2024
Description
Manufactured in compliance with ISO 594-1 standards, these fittings are cleaned, inspected and fully traceable. All parts meet tight dimensional tolerances for increased reliability and process purity.
Specifications
Specifications
5/32 in.
25 Pack