Learn More
Edmund Bühler GmbH™ Clamping Strip H
Holder with sponge rubber cover for fastening upright vessels.
77.35 EUR - 92.20 EUR
Specifications
Product Type | Clamping Strip |
---|---|
For Use With (Application) | Fastening upright vessels |
Includes | Sponge rubber cover |
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
10267461
|
Edmund Bühler GmbH™
0050 400 |
Shaker SM |
92.20 EUR
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10165841
|
Edmund Bühler GmbH™
0050 118 |
Combifix KS |
77.35 EUR
Each |
Estimated Shipment: 04-11-2024
Log in to see stock. |
|||||
Description
- Sponge rubber cover
- Fastens upright vessels
- For use with a variety of shakers
Specifications
Clamping Strip | |
Fastening upright vessels | |
Sponge rubber cover |