Learn More
Macherey-Nagel™ Chromofil™ Polyester Syringe Filter
Filter polar or nonpolar solvents. Chromofil™ PET Polyester Syringe Filter is hydrophilic for multipurpose applications and comes color coded.
Brand:  Macherey-Nagel™ 729021.400
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 10498213
  • 515.00 EUR / Pack of 400
Estimated Shipment: 04-07-2024
Description
  • Syringe filters with polyester membrane  
  • Color-coded 
  • Hydrophilic multipurpose membrane
  • For polar as well as nonpolar solvents
  • The HPLC filter, especially suited for mixtures of water and organic solvents 
  • For TOC/DOC determination
  • Not cytotoxic, does not inhibit the growth of microorganisms and higher cells
Specifications
Specifications
Yellow Top, Orange Bottom
0.2 μm
Hydrophilic
Syringe Filter
Polyester
Non Polar and Polar Solvent
400Pack
25 mm