missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlorotetracycline, MP Biomedicals™
Click to view available options
Quantity:
5 g
100 g
Specifications
Type | Chlorotetracycline |
Physical Form | Powder |
Formula Weight | 515.3 |
Color | Yellow |
Quantity | 5 g |
Chemical Name or Material | Chlorotetracycline |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'