Learn More
Eppendorf™ Centrifuge Adapters for Plate Rotors
For use with plate rotors.
139.00 EUR - 176.00 EUR
Specifications
Type | Adapter |
---|---|
Tube Type | PCR Plate |
For Use With (Equipment) | all MTP/DWP Rotor Buckets |
No. of Places | 4 |
No. per Pack | 2 Pack |
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
10446332
|
Eppendorf™
5825711009 |
96-Well PCR Plate |
176.00 EUR
Pack of 2 |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10314862
|
Eppendorf™
5825713001 |
384-Well PCR Plate |
139.00 EUR
Pack of 2 |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Available in either 96- or 384-well PCR plates
- Supplied in sets of 2
Specifications
Adapter | |
all MTP/DWP Rotor Buckets | |
2 Pack |
PCR Plate | |
4 |