Learn More
CD69 Armenian Hamster anti-Mouse, APC, Clone: H1.2F3, Proteintech
Armenian Hamster Monoclonal Antibody
Brand: Proteintech APC-65105-100UG
This item is not returnable.
View return policy
Description
The CD69 antibody from Proteintech is a armenian hamster monoclonal antibody generated with mouse dendritic epidermal t cell line y245 of mouse CD69. This antibody recognizes mouse antigen. The CD69 antibody has been validated for the following applications: FC analysis. This antibody is apc.
Specifications
CD69 | |
Monoclonal | |
0.2 mg/mL | |
Armenian Hamster | |
Primary | |
Mouse |
Flow Cytometry | |
H1.2F3 | |
APC | |
100 μg | |
12515 | |
IgG |