Learn More
Erlab™ Captair™ Work Surfaces
Designed for laboratories specializing in fields such as biology, botany, aerospace, electronics, pharmaceutical and cosmetics. Erlab™ Captair™ Work Surfaces are for use with Captair PCR work stations.
Brand:  Erlab™ I0115020002
Additional Details : Weight : 6.67800kg
 View more versions of this product
Product Code. 12361793
  • 747.30 EUR / Each
Estimated Shipment: 30-08-2024
Description
  • High chemical and mechanical resistance
  • Rounded corners to facilitate cleaning operations
  • Built in spill tray
Specifications
Specifications
For Captair™ Flex™ Model Midcap M321 and flow 321 fume hood
No. 1 Sealed Tempered Glass Retention Tray, Embedded Metal
Work Surface
Glass