Learn More
Erlab™ Captair Smart Filtering Fume Hood Mobicap
Erlab™ Captair Smart Filtering Fume Hood Mobicap offers a mobile, rolling bench for Captair Smart fume hoods.
Brand: Erlab™ I0562000201
Additional Details : Weight : 40.00000kg
Description
- For Captair Smart filtering fume hoods
- Equipped with 4 wheels and 2 self-locks
- Allows the fume hood to be safely moved
Specifications
Smart 481 Fume Hood | |
Without Work Surface | |
Metal |
890 mm | |
Mobile Rolling Cart Work Bench |