missing translation for 'onlineSavingsMsg'
Learn More

Calcium Carbonate, ≥99.0%, ACS Reagent, Honeywell Fluka™

Product Code. 15692420
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

ACS Reagent,≥99.0%, powder

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 471-34-1
Molecular Formula CCaO3
Molecular Weight (g/mol) 100.09
MDL Number MFCD00010906
InChI Key VTYYLEPIZMXCLO-UHFFFAOYSA-L
Synonym calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama
PubChem CID 10112
ChEBI CHEBI:3311
SMILES [Ca++].[O-]C([O-])=O

Specifications

Melting Point 825°C
Color White
Physical Form Powder
Quantity 2.5 kg
Linear Formula CaCO3
UN Number NONH for all modes of transport
Beilstein 8008338
IUPAC Name calcium;carbonate
Formula Weight 100.09g/mol
Percent Purity ≥99.0%
Grade ACS Reagent
Packaging Poly Bottle
Appearance Powder
Density 2.93g/mL at 25°C
Chemical Name or Material Calcium Carbonate(Caco3)
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'