Learn More
Buffer solutions, colour coded, BAKER ANALYZED™, J.T. Baker™
Click to view available options
Chemical Name or Material:
Phosphate
Phthalate
Quantity:
500 mL
1 L
Description
- Colour coded
- Ready-to-use pH buffers are manufactured in large lots to stringent specifications and utilize quality control procedures to reduce lot-to-lot variability.
- You can eliminate the time and expense of preparation and standardization of solutions, helping streamline high-volume analytical work in applications such as quality control.
- Solutions are available in both clear and colour-coded varieties.
- A certificate is provided to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
- Certifications: Each lot compared to NIST Standard Reference Buffer
Specifications
Specifications
Color | Red |
pH | 4.00 |
Quantity | 1 L |
Chemical Name or Material | Phthalate |