Learn More
Cole-Parmer™ Borosilicate Glass Media Bottles
![GREENER_CHOICE](/content/dam/fishersci/glyphs/green_glyph.png)
Click to view available options
Capacity (Metric):
250 mL
500 mL
Specifications
Capacity (Metric) | 250 mL |
Material | Borosilicate Glass |
No. per Case | 4Pack |
Quantity | 4Pack |
Closure Color | Orange |
Autoclavable | Autoclavable |
Product Type | Media Bottle |
Closure Material | Polypropylene |
Closure Type | Screw Cap |