Learn More
Robu™ Borosilicate Glass Crude Filter Crucibles
Brand: Robu™ 20602
Additional Details : Weight : 0.58500kg
Specifications
34mm | |
For crude fiber extraction | |
34mm | |
Crude Filter Crucible |
Borosilicate Glass | |
30 mL | |
60mm | |
2 |