Learn More
BRAND™ BLAUBRAND™ Borosilicate Glass Micro Burettes with PTFE Spindle
Constructed from borosilicate glass. BRAND™ Blaubrand™ Borosilicate Glass Micro Burettes with PTFE Spindle are calibrated to deliver contents that correspond to a volume as specified.
317.22 EUR - 346.80 EUR
Specifications
Material | Borosilicate Glass |
---|---|
Includes | Schellbach Stripes |
Stopcock | Lateral |
Stopcock Material | Glass |
Class | Class AS |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
10672441
|
BRAND™
24595 |
2 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11864050
|
BRAND™
24597 |
5 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10018500
|
BRAND™
24599 |
10 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Borosilicate glass construction (Boro 3.3. To DIN EN ISO 385)
- Calibrated to deliver (TD, Ex)
- Bang pattern
- Schellbach stripe
- Includes interchangeable needle-valve stopcock with PTFE spindle
- Stand included
- All BLAUBRAND™ burettes are delivered with batch number and enclosed one batch certificate per packing unit of the manufacturer
- On demand also available with individual certificate, USP Individual certificate or DAkkS calibration certificate
Specifications
Borosilicate Glass | |
Lateral | |
Class AS |
Schellbach Stripes | |
Glass | |
DIN EN ISO 385 |