Learn More
Agilent BioTek Liquid Handling Control Software
Offers dual mode control - runs protocols from PC or instrument keypad
Brand:  Agilent BioTek LHC2
Additional Details : Weight : 0.10000kg
Product Code. 15258768
  • 1910.00 EUR / Each
Estimated Shipment: 01-10-2024
Description
  • Intuitive StepWise protocol creation with an unlimited number of methods
  • Create and run protocols from PC or download to instrument's keypad
  • Automatic protocol validation for error-free assurance
  • Protocol parameters printed for safe record keeping and GLP compliance
  • Custom maintenance reminders for continued instrument performance in busy laboratories
  • Supports BioStack integrations for high throughput requirements
  • Security enabled protection for downloaded protocols
  • Multiple instruments controlled in parallel