Learn More
Agilent BioTek Absorbance Test Plate
Provides performance verification for microplate readers.
Brand:  Agilent BioTek 7260522
Additional Details : Weight : 0.20000kg
Product Code. 15810418
  • 1165.00 EUR / Each
Estimated Shipment: 14-08-2024
to see stock.
Includes:
  • Exact spectrophotometric calibration data for each neutral density filter at eight wavelengths: 405, 450, 490, 550, 620, 630, 690, and 750nm
  • Protective case
Description
Description
  • Tests alignment, accuracy, reproducibility and linearity
  • Conforms to the size and shape of a standard microplate to fit easily in a microplate reader
  • Ideal for monitoring an instrument's performance in compliance with regulatory requirements
Compliance
Uses NIST-traceable neutral density glass filters and precision machined blanking- and alignment-verification holes to confirm performance specifications.
Specifications
Specifications
405; 450; 490; 550; 620; 630; 690; 750 nm