Learn More
IBA Lifesciences BioLock
BioLock is a Biotin blocking solution for lysates and culture supernatants
Brand:  IBA Lifesciences 2-0205-050
Additional Details : Weight : 0.08100kg
 View more versions of this product
Product Code. 15643608
  • 213.00 EUR / 50mL
Estimated Shipment: 03-06-2024
Description
BioLock is a ready-to-use biotin blocking solution which contains the tetrameric biotin binding protein avidin. BioLock prevents biotin and biotinylated proteins from binding to Strep-Tactin™ or Strep-Tactin™XT, whereas binding of the Twin-Strep-tag™ and Strep-tag™II is not influenced.
Biotin and/or biotinylated proteins can possibly reduce the binding capacity of Strep-Tactin™ as well as Strep-Tactin™XT and are present in cell lysates and cell culture media, especially from mammalian or insect cells. Thus, if Twin-Strep-tag™ and Strep-tag™II fusion proteins from biotin-containing extracts or media are intended to be purified via Strep-Tactin™ or Strep-Tactin™XT affinity chromatography, biotin has to be masked by addition of BioLock prior to application onto the resin. 
Specifications
Specifications
50 mL
Activity: >70 U/ mL
Blocking Solution
Special Offers
For Research Use Only.