Learn More
Bel-Art™ SP Scienceware™ Magic Touch™ Iceware Ice Buckets
Lightweight ice buckets have excellent insulating properties for keeping temperature sensitive samples in an optimal environment
92.02 EUR - 121.86 EUR
Product Code | Brand | Capacity (Metric) | Description | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Description | Price | Quantity & Availability | ||||
12636435
|
Bel-Art™
M18848-4002 |
4 L | Black, With Lid Dia. x H: 23.5 x 20.1cm (9.25 x 7.94 in.) |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12616445
|
Bel-Art™
M18848-2001 |
2.5 L | Blue, With Lid Dia. x H: 22.2 x 12.0cm (8.75 x 4.75 in.) |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12696435
|
Bel-Art™
M18848-2002 |
2.5 L | Black, With Lid Dia. x H: 22.2 x 12.0cm (8.75 x 4.75 in.) |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
13402519
|
Bel-Art™
M18848-2201 |
2.5 L | Blue, Without Lid Dia. x H: 22.2 x 12.0cm (8.75 x 4.75 in.) |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12666435
|
Bel-Art™
M18848-4001 |
4 L | Blue, With Lid Dia. x H: 23.5 x 20.1cm (9.25 x 7.94 in.) |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Lightweight ice buckets have excellent insulating properties for keeping temperature sensitive samples in an optimal environment
- Temperature range: -196° to +100°C (-321° to +212°F)
- Use with ice, dry ice, dry ice alcohol slurries, or liquid nitrogen
- Nonsweating
- Impervious to moisture and odors
- Expanded polymer foam construction provides chemical resistance
- Unbreakable under normal use conditions
- Flared edge for easy carrying
- Lid can be cut to make openings for thermometers, cooling coils, flasks, etc.
- Available in two sizes: 2.5L buckets measure 22.2cm dia. at rim x 12.0cmH (8.75 x 4.75 in.); 4.0L buckets measure 23.5cm dia. at rim x 20.1cmH (9.25 x 7.9 in.)
- Round floating racks available separately for 1.5mL and 15mL tubes
Specifications
PVC |