Learn More
BD Disposable Syringes with Luer-Lok™ Tips
Unique integrated BD Luer-Lok™ tip syringe with clear barrel. BD Medical™ Disposable Syringes with Luer-Lok™ Tips are sterile and disposable.
416.00 EUR - 430.11 EUR
Specifications
Product Type | Syringe |
---|---|
Disposable | Yes |
Connection Type | Luer Lock |
Sterility | Sterile |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
10630694
|
BD
309628 |
1 mL |
416.00 EUR
Pack of 100 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15880305
|
BD
309628 |
1 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Unique integrated BD Luer-Lok™ tip syringe with clear barrel. BD Medical™ Disposable Syringes with Luer-Lok™ Tips are sterile and disposable.
- Sterile syringe for single use
- Without needle
- Clear barrel (polycarbonate or polypropylene) with bold scale markings
- Tapered plunger rod for ease of aspiration
- Positive plunger rod stop
- Added BD Luer-Lok™ thread for increased secure connection
- DNR (Latex) free
- Shelf life: 1825 days
Specifications
Syringe | |
Yes | |
Luer Lock | |
Sterile |