Learn More
BD BBL™ Taxo™ P Discs (Cartridge)
For presumptive identification of pneumococci, and are intended for use with pure cultures
Brand:  BD 231554
 View more versions of this product
Product Code. 17342381
  • 277.00 EUR / Pack of 10
Estimated Shipment: 20-11-2024
Description
  • 6mm discs made from high quality absorbent paper
  • Impregnated with approximately 5.0μg of ethylhydrocupreine hydrochloride per disc
Order Info
Cartridges of 50 discs.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Ethylhydrocupreine hydrochloride (optochin)
-20°C to +8°C
10 Cartridges
5μg
Differentiating Pneumococci
10 x 50 Discs
Special Offers
For in vitro Diagnostic Use.