Learn More
Berkshire™ BCR™ Telescoping Aluminum Handle
BCR™ Telescoping aluminum handle extends from 37-71 in. /95-180 cm.
Brand: Berkshire™ BCR.T2HANDLE.1
This item is not returnable.
View return policy
Description
- Telescopic handles available
- Anodized aluminum; lightweight and strong
- Autoclavable
- Telescopic handle extends from 37-71 in. /95-180 cm
Specifications
Gray | |
Autoclavable | |
ISO Class 3 to 7 |
95 to 180 cm | |
1 | |
Aluminum |