Learn More
Azlon™ Spouted HDPE Bucket
Spouted HDPE buckets for easy pouring and with moulded graduations
Brand:  Azlon BWX112
36.20 EUR valid until 2024-12-31
Use promo code "23259" to get your promotional price.
Additional Details : Weight : 0.55000kg
 View more versions of this product
Product Code. 12055562
  • 38.10 EUR / Each
Estimated Shipment: 18-06-2024
Description
    • Handle grip for easy lifting and carrying
    • Choice of white or black
    • Moulded graduation
Specifications
Specifications
Spouted Bucket
0.5 L
No
286 mm
1 Ea.
10.5 L
HDPE
White
283 mm