missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARHGEF10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00 EUR - 399.00 EUR
Specifications
Antigen | ARHGEF10 |
---|---|
Concentration | 0.13 mg/mL |
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16807973
|
Proteintech
11112-1-AP-20UL |
20 μL |
140.00 EUR
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16897963
|
Proteintech
11112-1-AP-150UL |
150 μL |
399.00 EUR
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene encodes a Rho guanine nucleotide exchange factor (GEF). Rho GEFs regulate the activity of small Rho GTPases by stimulating the exchange of guanine diphosphate (GDP) for guanine triphosphate (GTP) and may play a role in neural morphogenesis. Mutations in this gene are associated with slowed nerve conduction velocity (SNCV). Alternative splicing results in multiple transcript variants.Specifications
ARHGEF10 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Rat, Mouse, Human | |
O15013, Q8C033 | |
234094, 306618, 9639 | |
ARHGEF10 Fusion Protein Ag1605 | |
Primary | |
-20°C | |
ARHGEF10 |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
ARHGEF10, GEF10, KIAA0294 | |
ARHGEF10 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |