Learn More
BINDER™ Arabidopsis Illumination
Arabidopsis Illumination - Illumination cassettes equipped with Arabidopsis fluorescent tubes instead of daylight fluorescent tubes
Brand: BINDER™ 8012-1499
Additional Details : Weight : 2.10000kg
Beschreibung
- 17 W or 25 W option model
- For use with a range of growth chambers
Spezifikation
Arabidopsis illumination - Illumination cassettes equipped with Arabidopsis fluorescent tubes instead of daylight fluorescent tubes | |
BINDER Chambers KBWF 720 | |
Fluorescent Light | |
25 W |